Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fzd5
Entrez Gene ID
100762351
Gene Name
frizzled class receptor 5
Gene Aliases
Frizzled-5, I79_014467
Chromosome Location
Chr.Un|NW_003614305: 509926 - 514402 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506923.2 XP_003506971.1 2 - 3 2006 107

View Full Product Details