Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kbtbd2
Entrez Gene ID
100768350
Gene Name
kelch repeat and BTB (POZ) domain containing 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614101: 334782 - 350319 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504900.2 XP_003504948.1 3 - 4 677 102
GenBank mRNA JI872608.1 - 2 - 3 694 102
JP065993.1 - 2 - 3 694 102

View Full Product Details