Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcp2
Entrez Gene ID
100768267
Gene Name
Purkinje cell protein 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614420: 511641 - 513863 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507746.2 XP_003507794.2 1 - 2 52 95

View Full Product Details