Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slfn14
Entrez Gene ID
100752549
Gene Name
schlafen family member 14
Gene Aliases
I79_019785
Chromosome Location
Chr.Un|NW_003615071: 300565 - 308945 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511309.1 XP_003511357.1 3 - 4 1871 101

View Full Product Details