Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nkx2-8
Entrez Gene ID
100774048
Gene Name
NK2 homeobox 8
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615762: 108761 - 110075 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513155.1 XP_003513203.1 1 - 2 143 84

View Full Product Details