Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cnga4
Entrez Gene ID
100753462
Gene Name
cyclic nucleotide gated channel alpha 4
Gene Aliases
I79_022089
Chromosome Location
Chr.Un|NW_003615765: 59083 - 64873 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513168.2 XP_003513216.1 4 - 5 440 95

View Full Product Details