Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prdm8
Entrez Gene ID
100763807
Gene Name
PR domain containing 8
Gene Aliases
I79_017275
Chromosome Location
Chr.Un|NW_003614663: 326358 - 330709 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649769.1 XP_007647959.1 2 - 3 800 71

View Full Product Details