Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
March1
Entrez Gene ID
100753515
Gene Name
membrane-associated ring finger (C3HC4) 1, E3 ubiquitin protein ligase
Gene Aliases
I79_010978
Chromosome Location
Chr.Un|NW_003614020: 605244 - 919341 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644539.1 XP_007642729.1 5 - 6 608 76

View Full Product Details