Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hcn4
Entrez Gene ID
100758754
Gene Name
hyperpolarization activated cyclic nucleotide-gated potassium channel 4
Gene Aliases
I79_008977
Chromosome Location
Chr.Un|NW_003613894: 1087020 - 1125738 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643027.1 XP_007641217.1 2 - 3 556 68

View Full Product Details