Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dspp
Entrez Gene ID
100754381
Gene Name
dentin sialophosphoprotein
Gene Aliases
I79_025469
Chromosome Location
Chr.Un|NW_003619489: 12535 - 16873 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515762.1 XP_003515810.1 1 - 2 54 103

View Full Product Details