Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h12d
Entrez Gene ID
100759590
Gene Name
zinc finger CCCH-type containing 12D
Gene Aliases
I79_004914
Chromosome Location
Chr.Un|NW_003613710: 435882 - 460064 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498945.2 XP_003498993.1 2 - 3 456 116

View Full Product Details