Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fstl3
Entrez Gene ID
103160351
Gene Name
follistatin-like 3 (secreted glycoprotein)
Gene Aliases
I79_009196
Chromosome Location
Chr.Un|NW_003613904: 552369 - 566008 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643245.1 XP_007641435.1 2 - 3 264 71

View Full Product Details