Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pvrig
Entrez Gene ID
103161366
Gene Name
poliovirus receptor related immunoglobulin domain containing
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614875: 396998 - 401040 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651044.1 XP_007649234.1 5 - 6 925 59

View Full Product Details