Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb42
Entrez Gene ID
100752108
Gene Name
zinc finger and BTB domain containing 42
Gene Aliases
I79_004713
Chromosome Location
Chr.Un|NW_003613702: 2229036 - 2239619 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007633192.1 XP_007631382.1 2 - 3 308 94

View Full Product Details