Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr49
Entrez Gene ID
100760310
Gene Name
WD repeat domain 49
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614182: 703232 - 847803 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646212.1 XP_007644402.1 11 - 12 1986 70

View Full Product Details