Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plscr3
Entrez Gene ID
100759961
Gene Name
phospholipid scramblase 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613643: 1904390 - 1908716 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007608471.1 XP_007606661.1 2 - 3 575 104

View Full Product Details