Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sntn
Entrez Gene ID
100754229
Gene Name
sentan, cilia apical structure protein
Gene Aliases
I79_010541, Sentan
Chromosome Location
Chr.Un|NW_003613987: 898692 - 916283 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503673.2 XP_003503721.2 1 - 2 110 97

View Full Product Details