Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pla2g4c
Entrez Gene ID
100754559
Gene Name
phospholipase A2, group IVC (cytosolic, calcium-independent)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615727: 181003 - 207358 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513105.1 XP_003513153.1 9 - 10 1115 94

View Full Product Details