Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tsen2
Entrez Gene ID
100768441
Gene Name
TSEN2 tRNA splicing endonuclease subunit
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614360: 21196 - 74578 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647912.1 XP_007646102.1 3 - 4 5123 65
XM_007647913.1 XP_007646103.1 3 - 4 4278 65
XM_007647914.1 XP_007646104.1 3 - 4 3541 65
XM_007647915.1 XP_007646105.1 3 - 4 3578 65
XM_007647916.1 XP_007646106.1 2 - 3 5596 65
XM_007647917.1 XP_007646107.1 3 - 4 3462 65
XM_007647918.1 XP_007646108.1 4 - 5 1628 65
XM_007647919.1 XP_007646109.1 3 - 4 1549 65
XM_007647920.1 XP_007646110.1 2 - 3 1287 65
XM_007647921.1 XP_007646111.1 2 - 3 296 65
XM_007647922.1 XP_007646112.1 2 - 3 540 65
XM_007647927.1 XP_007646117.1 2 - 3 5379 65

View Full Product Details