Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tro
Entrez Gene ID
100759986
Gene Name
trophinin
Gene Aliases
I79_006415
Chromosome Location
Chr.Un|NW_003613780: 480739 - 489925 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640914.1 XP_007639104.1 1 - 2 40 112

View Full Product Details