Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrpl18
Entrez Gene ID
100760587
Gene Name
mitochondrial ribosomal protein L18
Gene Aliases
I79_009787
Chromosome Location
Chr.Un|NW_003613938: 56953 - 60972 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503059.2 XP_003503107.1 1 - 2 640 77
XM_007643682.1 XP_007641872.1 1 - 2 640 77
GenBank mRNA JP061042.1 - 1 - 2 216 77

View Full Product Details