Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fgf21
Entrez Gene ID
100757339
Gene Name
fibroblast growth factor 21
Gene Aliases
I79_016636
Chromosome Location
Chr.Un|NW_003614570: 137373 - 138701 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508678.2 XP_003508726.1 2 - 3 435 76

View Full Product Details