Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Efhd1
Entrez Gene ID
100765009
Gene Name
EF-hand domain family, member D1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615857: 118105 - 162408 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513317.1 XP_003513365.1 4 - 5 570 103

View Full Product Details