Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Narg2
Entrez Gene ID
100759663
Gene Name
NMDA receptor regulated 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616066: 63070 - 100926 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513659.2 XP_003513707.1 4 - 5 617 134
XM_007653918.1 XP_007652108.1 4 - 5 547 134
XM_007653919.1 XP_007652109.1 4 - 5 714 134
GenBank mRNA JI891276.1 - 4 - 5 574 134
JP044394.1 - 4 - 5 574 134

View Full Product Details