Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Samd12
Entrez Gene ID
100753622
Gene Name
sterile alpha motif domain containing 12
Gene Aliases
I79_007334
Chromosome Location
Chr.Un|NW_003613825: 563656 - 944637 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641717.1 XP_007639907.1 1 - 2 220 67

View Full Product Details