Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slitrk1
Entrez Gene ID
100754536
Gene Name
SLIT and NTRK-like family, member 1
Gene Aliases
I79_015968
Chromosome Location
Chr.Un|NW_003614487: 474303 - 476908 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508135.2 XP_003508183.1 1 - 2 63 110

View Full Product Details