Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tdrd6
Entrez Gene ID
100759643
Gene Name
tudor domain containing 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614784: 474254 - 490427 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509859.1 XP_003509907.1 3 - 4 5824 108

View Full Product Details