Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mei4
Entrez Gene ID
100765662
Gene Name
meiosis-specific 4 homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614982: 58900 - 221869 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510873.2 XP_003510921.2 2 - 3 302 71
XM_007651459.1 XP_007649649.1 2 - 3 291 71

View Full Product Details