Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Iqcj
Entrez Gene ID
103160084
Gene Name
IQ motif containing J
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614175: 366815 - 388435 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646144.1 XP_007644334.1 3 - 4 224 75

View Full Product Details