Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pram1
Entrez Gene ID
100755895
Gene Name
PML-RARA regulated adaptor molecule 1
Gene Aliases
I79_018808
Chromosome Location
Chr.Un|NW_003614895: 213838 - 219099 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651134.1 XP_007649324.1 6 - 7 1965 105

View Full Product Details