Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Exosc4
Entrez Gene ID
100766618
Gene Name
exosome component 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614411: 432412 - 434234 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507675.2 XP_003507723.1 2 - 3 458 65
GenBank mRNA JP041945.1 - 2 - 3 445 65

View Full Product Details