Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tagln3
Entrez Gene ID
100764031
Gene Name
transgelin 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613667: 511960 - 525530 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497847.2 XP_003497895.1 1 - 2 181 99

View Full Product Details