Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam71f2
Entrez Gene ID
100750641
Gene Name
family with sequence similarity 71, member F2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613652: 1464957 - 1475074 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007613390.1 XP_007611580.1 3 - 4 705 99
XM_007613394.1 XP_007611584.1 4 - 5 749 99

View Full Product Details