Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim41
Entrez Gene ID
100767959
Gene Name
tripartite motif containing 41
Gene Aliases
I79_011011
Chromosome Location
Chr.Un|NW_003614027: 873766 - 883527 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644569.1 XP_007642759.1 2 - 3 551 75

View Full Product Details