Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Taf13
Entrez Gene ID
100766831
Gene Name
TAF13 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 18kDa
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615469: 172303 - 181102 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512482.2 XP_003512530.1 3 - 4 229 82

View Full Product Details