Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ms4a14
Entrez Gene ID
100754368
Gene Name
membrane-spanning 4-domains, subfamily A, member 14
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616909: 18707 - 40095 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514750.1 XP_003514798.1 1 - 2 135 69

View Full Product Details