Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slfnl1
Entrez Gene ID
100757695
Gene Name
schlafen-like 1
Gene Aliases
I79_008247
Chromosome Location
Chr.Un|NW_003613862: 664729 - 668451 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642471.1 XP_007640661.1 4 - 5 1356 78

View Full Product Details