Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Laptm5
Entrez Gene ID
100766375
Gene Name
lysosomal protein transmembrane 5
Gene Aliases
I79_004164
Chromosome Location
Chr.Un|NW_003613689: 820414 - 841638 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498363.2 XP_003498411.1 2 - 3 180 64

View Full Product Details