Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtfp1
Entrez Gene ID
100755489
Gene Name
mitochondrial fission process 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614628: 188861 - 193382 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509012.2 XP_003509060.1 1 - 2 1325 75
XM_007649642.1 XP_007647832.1 2 - 3 682 75
XM_007649643.1 XP_007647833.1 2 - 3 905 75
XM_007649644.1 XP_007647834.1 2 - 3 697 75
XM_007649645.1 XP_007647835.1 2 - 3 595 75
XM_007649646.1 XP_007647836.1 2 - 3 818 75
XM_007649647.1 XP_007647837.1 2 - 3 610 75

View Full Product Details