Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb9
Entrez Gene ID
100773744
Gene Name
zinc finger and BTB domain containing 9
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614972: 267916 - 274093 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510826.2 XP_003510874.1 1 - 2 1854 98

View Full Product Details