Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdzd3
Entrez Gene ID
100756376
Gene Name
PDZ domain containing 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615154: 105787 - 109577 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511555.2 XP_003511603.1 8 - 9 1057 80

View Full Product Details