Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tssk4
Entrez Gene ID
100758937
Gene Name
testis-specific serine kinase 4
Gene Aliases
I79_008540
Chromosome Location
Chr.Un|NW_003613875: 1066138 - 1077538 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502057.2 XP_003502105.1 2 - 3 1280 59

View Full Product Details