Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sdc4
Entrez Gene ID
100760006
Gene Name
syndecan 4
Gene Aliases
Syndecan-4
Chromosome Location
Chr.Un|NW_003614077: 846868 - 866029 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504581.2 XP_003504629.1 1 - 2 117 79
GenBank mRNA JI865108.1 - 1 - 2 95 79
JP062945.1 - 1 - 2 99 79

View Full Product Details