Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim44
Entrez Gene ID
100757190
Gene Name
tripartite motif containing 44
Gene Aliases
I79_003047
Chromosome Location
Chr.Un|NW_003613649: 2361005 - 2455531 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007612568.1 XP_007610758.1 3 - 4 757 84
GenBank mRNA JP047629.1 - 3 - 4 349 84

View Full Product Details