Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam89a
Entrez Gene ID
100772991
Gene Name
family with sequence similarity 89, member A
Gene Aliases
I79_002297
Chromosome Location
Chr.Un|NW_003613636: 2293820 - 2360609 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496834.1 XP_003496882.1 7 - 8 1297 104

View Full Product Details