Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr6
Entrez Gene ID
100765288
Gene Name
WD repeat domain 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615379: 113223 - 117487 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652651.1 XP_007650841.1 1 - 2 2582 78
GenBank mRNA JP041824.1 - 1 - 2 1247 78

View Full Product Details