Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asf1a
Entrez Gene ID
103163586
Gene Name
anti-silencing function 1A histone chaperone
Gene Aliases
I79_018925
Chromosome Location
Chr.Un|NW_003614914: 30226 - 36959 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651209.1 XP_007649399.1 2 - 3 421 95
GenBank mRNA JI877335.1 - 2 - 3 421 95

View Full Product Details