Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Irx5
Entrez Gene ID
100773873
Gene Name
iroquois homeobox 5
Gene Aliases
I79_003733
Chromosome Location
Chr.Un|NW_003613676: 2255043 - 2258208 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498019.2 XP_003498067.1 2 - 3 604 73

View Full Product Details