Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100755558
Entrez Gene ID
100755558
Gene Name
chromosome unknown open reading frame, human C1orf85
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613923: 1170131 - 1173602 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502912.2 XP_003502960.1 4 - 5 845 86
GenBank mRNA JI893402.1 - 4 - 5 853 86
JP043948.1 - 4 - 5 853 86

View Full Product Details