Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fsip2
Entrez Gene ID
100754113
Gene Name
fibrous sheath interacting protein 2
Gene Aliases
I79_010180
Chromosome Location
Chr.Un|NW_003613964: 1112877 - 1196514 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503397.1 XP_003503445.1 8 - 9 991 153

View Full Product Details